Learn More
Thermo Scientific™ Abgene™ 96 Well 1.2mL Polypropylene DeepWell™ Sample Processing & Storage Plate
Optimize suspension cell culture with these U-bottomed 1.2 mL storage plates.
268.00 € - 286.00 €
Specifications
Barcode | Custom Barcoding Available |
---|---|
Certifications/Compliance | Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant |
Format | 96-well |
Description | 96-well Polypropylene Sample Processing & Storage Plates |
Array | 8 x 12 |
Product Code | Brand | Well Shape | Packaging | Fit for Purpose | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Well Shape | Packaging | Fit for Purpose | Price | Quantity & Availability | |||
10243223
|
Thermo Scientific™
AB-1127 |
Square, Round Bottom (U-well) | Individually Wrapped | PCR Ready |
271.00 €
Pack of 50 |
In Stock
Log in to see stock. |
|||
12114172
|
Thermo Scientific™
AB-0564 |
Round | Individually Wrapped | PCR Ready |
268.00 €
Pack of 50 |
In Stock
Log in to see stock. |
|||
10544384
|
Thermo Scientific™
AB-0787 |
Round | Bulk | PCR Ready |
286.00 €
Pack of 50 |
In Stock
Log in to see stock. |
|||
10364463
|
Thermo Scientific™
AB-0564/K |
Round | Individually Wrapped | PCR Ready |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Thermo Scientific™ Abgene™ polypropylene microplates and deepwell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use. Abgene microtiter plates are manufactured to exacting specifications in our Class 100,000 clean room ISO 9001 conditions using high quality medical grade virgin polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and deepwell plates are compatible with a variety of automated liquid handling for high throughput workflows. The polypropylene storage plates are offered with spindle, pyramidal, U and V bottom wells along with a wide range of volumes to accommodate most applications.
Features
- Round well shape designed for optimal sample recovery or square shape to maximize sample volume within ANSI footprint design
- Each well has an independent sealing rim to prevent cross-contamination
- U-bottomed wells ideally suited for sample resuspension
- Polypropylene (AB-1068, polystyrene)
- Resistant to DMSO (100%); Ethanol (100%); Isopropanol (100%)
- Suitable for Autoclaving (15 minutes at 121°C) or Gamma Irradiation
- Sealing options: Adhesive Seals, Heat Seals, Storage Plate Caps and Cap Strips, and Storage Plate Sealing Mats
- Cleanroom manufactured
- ANSI-format for compatibility with automated systems
Specifications
Custom Barcoding Available | |
96-well | |
8 x 12 | |
Abgene | |
ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers | |
1200 μL | |
Cleanroom | |
Yes | |
50 | |
Yes | |
ANSI/ SLAS Standard |
Certified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant | |
96-well Polypropylene Sample Processing & Storage Plates | |
Low binding PP | |
96-well plate | |
Polypropylene | |
96 | |
Non-treated | |
Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation | |
For Research Use Only. Not for use in diagnostic procedures. | |
PCR Ready |