Learn More
Cytiva AC 16 Flow Adapter
Upgrade fixed volume C columns by adding flow adapters (which keep bed height constant and protect from insoluble particles) and a thermostatic jacket. Cytiva AC 16 Flow Adapters are available in different sizes for different column diameters.
Brand: Cytiva 19-5109-01
Code : GE
Additional Details : Weight : 0.18200kg
Description
- Three adjustable flow adapters are available for the C column series (AC 10, AC 16, and AC 26),
- Enabling optimal bed heights for each application, as are thermostatic jackets for precise temperature control
Specifications
Fluoro Rubber, Fluro Carbon Rubber (FPM), Polyamide, Polyethylene, Polypropylene, Stainless Steel | |
AC 16 Flow Adapter | |
1 Pack |