missing translation for 'onlineSavingsMsg'
Learn More

Merck Accessories for Analytical Filter Holders

Accessories for filter holders for sample preparation.

Brand:  Merck XX2014718

Additional Details : Weight : 0.81710kg

 View more versions of this product

Product Code. 15844176

  • 208.00 € / Each
Estimated Shipment: 15-07-2024
to see stock.
Add to basket

Description

Description

Accessories for filter holders for sample preparation.

  • Not Autoclaved
Specifications

Specifications

Not Autoclavable
Millipore 25 mm Analytical Stainless Steel Filter Holder Kit
Non-sterile
Stopper, No 8
9.5 mm hole diameter
Silicone with parylene coating
5/Pack
47 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers