missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Accessories and Upgrade Kits for Varioskan LUX Multimode Microplate Reader
Various accessories for use with the Varioskan™ LUX Multimode Microplate Reader.
36.96 € - 15967.00 €
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
15330787
|
Thermo Scientific™
N16443 |
Storage holder for plate adapters |
50.25 €
Each |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15340787
|
Thermo Scientific™
N16484 |
Emission filter pick-up tool |
36.96 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12433458
|
Thermo Scientific™
2805690 |
Aspirate tube assembly for a dispenser |
107.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12439757
|
Thermo Scientific™
F42550 |
Emission Filter |
910.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11599479
|
Thermo Scientific™
N02339 |
Robotic plate tray adapter for plates without lid |
289.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12472238
|
Thermo Scientific™
N02690 |
384-well adapter for plate without lid |
576.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12871886
|
Thermo Scientific™
N02691 |
384-well adapter for plate with lid |
576.00 €
Each |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12492238
|
Thermo Scientific™
N02692 |
96-well adapter for plate without lid |
578.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11905484
|
Thermo Scientific™
N02693 |
96-well adapter for plate with lid |
566.00 €
Each |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11589309
|
Thermo Scientific™
N02696 |
6-48-well adapter for plate without lid |
289.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12432248
|
Thermo Scientific™
N03078 |
Universal plate tray for plates without lid |
3835.00 €
Each |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12442248
|
Thermo Scientific™
N03079 |
Robotic plate tray for plates with and without lids |
1628.00 €
Each |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12452248
|
Thermo Scientific™
N03395 |
96-well adapter for PCR plate without lid |
982.00 €
Each |
Estimated Shipment: 26-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12472248
|
Thermo Scientific™
N06210 |
1536-well adapter for 10 mm plate without lid |
982.00 €
Each |
In Stock
Log in to see stock. |
|||||
12492248
|
Thermo Scientific™
SP-00093 |
Dispenser syringe, 5 mL |
560.00 €
Each |
Estimated Shipment: 17-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12402258
|
Thermo Scientific™
SP-00094 |
Dispensing tube assembly for a dispenser |
490.00 €
Each |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12412258
|
Thermo Scientific™
SP-00096 |
Dispenser syringe, 1 mL |
542.00 €
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17198238
|
Thermo Scientific™
SP-00720 |
Microplate package for Varioskan LUX IQ/OQ |
325.00 €
Each |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17954312
|
Thermo Scientific™
N16308S |
Liquid protection tray |
74.80 €
Each |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17972051
|
Thermo Scientific™
VLUPLA06 |
Upgrade kit for luminometry and AlphaScreen |
11975.00 €
Each |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17982081
|
Thermo Scientific™
VLUPLT07 |
Upgrade kit for luminometry and TRF |
11975.00 €
Each |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17932161
|
Thermo Scientific™
VLUPLAT08 |
Upgrade kit for luminometry, AlphaScreen and TRF |
15967.00 €
Each |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17982051
|
Thermo Scientific™
VLUPD209 |
Upgrade kit for two dispensers |
7984.00 €
Each |
Estimated Shipment: 13-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Thermo Scientific™ Varioskan™ LUX multimode microplate reader is a versatile tool for busy labs equipped with a flexible range of measurement technologies including Absorbance, Fluorescence Intensity, Luminescence, AlphaScreen, and Time-Resolved Fluorescence. You can configure the instrument to your needs, and when your research focus changes, just upgrade the instrument with service installable upgrade kits. Various plate adapters and other accessories helps you to optimize your assays.
Varioskan LUX is a modular instrument, you can upgrade it with missing features later.- Installation of the upgrade kits must be carried out by trained and authorized service personnel.
- For TRF and/or AlphaScreen technology upgrades, the instrument must be equipped with a luminescence module.
- TRF and/or AlphaScreen technology upgrades include excitation filters but not emission filters. Please order emission filters separately.
- Please contact your local representative to discuss about the instrument upgrade option
- Adapters adjust the plates to the optimum height for measuring and dispensing.
- Choose the adapter based on the plate format in use and whether the lid is needed or not.
- The universal plate tray is for basic use and it is compatible with all plate formats (6- to 1536-well plates).
- A robotic plate tray is for automated use with robots and is compatible with 96- to 1536-well plate formats.
If the dispensers are in frequent use, it is recommended to replace the aspirate and dispensing tubings once a year.
Specifications
Varioskan™ |