Learn More
BD Accuri™ Bacteriostatic Concentrate Solution
Prevents the growth of bacteria in the sheath fluid of the Accuri™ C6 Flow Cytometer. BD™ Accuri™ Bacteriostatic Concentrate Solution can be used to treat 10L of sheath fluid. Packaged in a pack of ten 8mL vials.
Brand: BD 653156
Code : NEW
Additional Details : CAS Number : 7681-49-4 Weight : 0.10000kg Transport : UN number : 1760 Chem class : 8 Pack group : II
Description
- Application: Flow cytometry (particularly BD Accuri™ C6 Flow Cytometer)
- Added to 10L deionized water to prevent the growth or growth of bacteria in the sheath fluid
- Lot number and expiration date listed on label of each vial
- For research only – not for therapeutic or diagnostic use
Specifications
Ten 8mL vials | |
Liquid | |
Colorless, clear |
Store in cool, dry conditions in well sealed receptacles. | |
Bacteriostatic Agent | |
8 mL |