missing translation for 'onlineSavingsMsg'
Learn More

Ansell AccuTech™ 91-325 Series Natural Rubber Latex Splash Gloves

Catalog Number 15265183
Click to view available options
Size:
6
6.5
7
7.5
8
8.5
9
This item is not returnable. View return policy
This item is not returnable. View return policy

Provides optimal solutions for controlled and sterile environments, respectively. Ansell Edmont™ AccuTech™ 91-325 Series Natural Rubber Latex Splash Gloves are made with a specific natural rubber formulation that offers a high level of comfortable, sensitive, precise handling.

  • With textured fingertips
  • Excellent technical properties
  • Low allergenic content minimizes the risk of latex-associated allergies for the wearer
  • Ideal for biotechnologies, medical device manufacturing, laboratory analysis, pharmaceuticals and the flat panel industry
Cleanroom Class Class 100 (ISO 5)
Color Natural
Latex Free No
Material Latex
Product Type Cleanroom Gloves
Size 6
Thickness (Metric) 9.8 mm
For Use With (Application) For biotechnologies, analytics testing, blistering operations, weighing and dispensing etc.
Glove Cuff Color Natural
Glove Cuff Style Straight
Quantity 200 Pairs
Series 91-325
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'