missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Glacial, BAKER INSTRA-ANALYZED Plus, J.T. Baker™

Catalog Number 15548594
Click to view available options
Quantity:
500 mL
1 L
2.5 L
This item is not returnable. View return policy
64-19-7
C2H4O2
60.05
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
176
CHEBI:15366
acetic acid
CC(O)=O
This item is not returnable. View return policy
64-19-7
C2H4O2
60.05
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
176
CHEBI:15366
acetic acid
CC(O)=O

Analyzed for up to 64 metals in the low parts-per-billion (ppb) range, ensuring low background interference for conducting critical trace metal analysis.

BAKER INSTRA-ANALYZED™ Plus acids allow for testing of more trace metals with tighter specifications.

  • Quality assured for up to 64 trace metals tested to low parts-per-billion (ppb) levels
  • Lot to lot consistency ensures reliable results between tests
  • Space-saving HDPE packaging with low metallic extractables
  • Enhanced safety with tamper-evident caps
  • Environmentally-friendly recyclable HDPE bottles
  • Strict protocols in an ISO facility ensure quality in manufacturing
pH 2.4 (1.0M aq. soln.)
Melting Point 16.4°C
Boiling Point 117.9°C
Color Colorless
Linear Formula CH3CO2H
Solubility Information Miscible in water
Formula Weight 60.05g/mol
Density 1.05g/mL at 20°C
Percent Purity 99% (min.)
Grade For use in Trace Metal Analysis
Quantity 1 L
Explosion Limit 4 - 19.9 Vol %
Flash Point 39.4°C
Ignition Point 426°C
Odor Pungent, vinegar-like
Packaging HDPE
Physical Form Liquid
Vapor Density 2.1
Vapor Pressure 2.093 kPa
Assay w/w
Chemical Name or Material Acetic acid, glacial
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'