missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid Glacial, USP, Macron Fine Chemicals™

Catalog Number p-7138030
83.51 EUR valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
0.5 L
2.5 L
Packaging:
Clear bottle
Clear glass bottle
This item is not returnable. View return policy
64-19-7
C2H4O2
60.05
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
176
CHEBI:15366
acetic acid
CC(O)=O
This item is not returnable. View return policy
64-19-7
C2H4O2
60.05
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
176
CHEBI:15366
acetic acid
CC(O)=O

Conforms to the requirements of the United States Pharmacopeia (USP).

  • Meets the requirements necessary for use in pharmaceutical processing and prescription compounding
  • Product is regulated by the Food and Drug Administration
  • Manufactured under current Good Manufacturing Practices (cGMP)
pH 2.5 (at 20°C)
Melting Point 16.6°C
Boiling Point 118°C
Color Colorless
Appearance Clear
Assay Percent Range 99.7 to 100.5%
Linear Formula CH3COOH
Solubility Information Fully miscible in water
Formula Weight 60.05g/mol
Grade USP
Density 1.05g/cm3 at 20°C
Quantity 0.5 L
Flash Point 39 to 40°C
Packaging Clear glass bottle
Physical Form Liquid
Chemical Name or Material Glacial Acetic Acid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P370+P378-In case of fire: Use to extinguish.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'