missing translation for 'onlineSavingsMsg'
Learn More

Acetic Acid, Optima™ LC/MS Grade, Fisher Chemical™

Catalog Number p-7072847
Click to view available options
Quantity:
1 x 1 mL Ampule
10 x 1 mL Ampule
50 x 1 mL Ampule
This item is not returnable. View return policy
64-19-7
C2H4O2
60.05
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
CC(O)=O
This item is not returnable. View return policy
64-19-7
C2H4O2
60.05
MFCD00036152
QTBSBXVTEAMEQO-UHFFFAOYSA-N
acetic acid
CC(O)=O

C2H4O2, CAS Number-64-19-7, 50mL, 117 deg.C, Colorless, 60.05g/mol, 16 deg.C, 36152, 2.5, Liquid, 2.1, 1.52kPa at 20 deg.C

TRUSTED_SUSTAINABILITY
pH 2.5
Melting Point 16°C
Boiling Point 117°C
Color Colorless
Formula Weight 60.05g/mol
Quantity 1 x 1 mL Ampule
Physical Form Liquid
Vapor Density 2.1
Vapor Pressure 1.52kPa at 20°C
compliance-icons compliance-icons
Product Identifier
  • Acetic acid, Optima LCMS
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Flammable liquid Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'