missing translation for 'onlineSavingsMsg'
Learn More

Acetic anhydride, 99+%, Thermo Scientific Chemicals

Catalog Number p-3737907
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic Drum
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
108-24-7
C4H6O3
102.089
MFCD00008705
WFDIJRYMOXRFFG-UHFFFAOYSA-N
acetic anhydride, acetanhydride, acetic acid, anhydride, acetic oxide, acetyl oxide, ethanoic anhydride, acetyl ether, acetyl anhydride, acetic acid anhydride, anhydride acetique
7918
CHEBI:36610
acetyl acetate
CC(=O)OC(=O)C
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
108-24-7
C4H6O3
102.089
MFCD00008705
WFDIJRYMOXRFFG-UHFFFAOYSA-N
acetic anhydride, acetanhydride, acetic acid, anhydride, acetic oxide, acetyl oxide, ethanoic anhydride, acetyl ether, acetyl anhydride, acetic acid anhydride, anhydride acetique
7918
CHEBI:36610
acetyl acetate
CC(=O)OC(=O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Acylation reagent for analysis
TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Color Yellow
Density 1.0870g/mL
Assay Percent Range 99+%
Packaging Glass bottle
Linear Formula (CH3CO)2O
Quantity 500 mL
Beilstein 02, 166
Residue after Evaporation 0.003 % max.
Fieser 01,3; 02,7; 05,3; 06,1; 07,1; 08,1; 09,1; 11,1; 12,1; 13,93; 14,150; 15,1
Merck Index 15, 55
Formula Weight 102.09
Percent Purity ≥99.0%
Grade Analysis
Physical Form Liquid
Chemical Name or Material Acetic anhydride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Acetic anhydride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'