missing translation for 'onlineSavingsMsg'
Learn More

Acetoacetic acid sodium salt, MedChemExpress

Product Code. 16434648
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16434648

Brand: Medchem Express HY112540B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Acetoacetic acid sodium salt is a metabolite of non-esterified fatty acids, involved in the development of human diabetes. Acetoacetic acid sodium salt induces oxidative stress to inhibit the assembly of very low density lipoprotein in bovine hepatocytes.

Acetoacetic acid sodium salt is a metabolite of non-esterified fatty acids, involved in the development of human diabetes. Acetoacetic acid sodium salt induces oxidative stress to inhibit the assembly of very low density lipoprotein in bovine hepatocytes.

Specifications

Chemical Name or Material Acetoacetic acid sodium salt
Physical Form Solid
CAS 623-58-5
Color Pink
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C4H5NaO3
For Use With (Application) Metabolism-sugar/lipid metabolism
Solubility Information H2O : 50 mg/mL (403.00 mM; Need ultrasonic)
SMILES CC(CC(O[Na])=O)=O
Molecular Weight (g/mol) 124.07
Formula Weight 124.07
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.