missing translation for 'onlineSavingsMsg'
Learn More

Acetone, 99.8%, for HPLC, Thermo Scientific Chemicals

Catalog Number p-217171
52.65 EUR valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 L
2.5 L
67-64-1
C3H6O
58.08
MFCD00008765
CSCPPACGZOOCGX-UHFFFAOYSA-N
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
180
CHEBI:15347
propan-2-one
CC(C)=O
67-64-1
C3H6O
58.08
MFCD00008765
CSCPPACGZOOCGX-UHFFFAOYSA-N
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
180
CHEBI:15347
propan-2-one
CC(C)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
TRUSTED_SUSTAINABILITY
Melting Point -95.0°C
Absorbance (1 cm cell vs HPLC−grade water), (1 cm cell vs HPLC−grade water) at 0nm, 0.005 max. at 350nm, 0.005 max. at 360nm, 0.005 max. at 370nm, 0.09 max. at 340nm, 0.3 max. at 335nm, 1 max. at 330nm
CAS Min % 99.75
Acidity ≤0.0003meq/g
CAS Max % 100.0
Alkalinity ≤0.0003meq/g
Decomposition Information >4°C
Density 0.7900g/mL
Boiling Point 56.0°C
Quantity 1 L
Flash Point −20°C
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3585
Assay Percent Range 99.8%
Residue after Evaporation ≤0.0002%
Specific Gravity 0.79
Linear Formula CH3COCH3
Vapor Pressure Vapor Pressure: 247mbar at 20°C
UN Number 1090; Colorless
Viscosity 0.32 mPa.s (20°C)
Beilstein 01, IV, 3180
Water ≤0.1%
Fieser 02,13; 03,4; 06,9
Merck Index 15, 65
Solubility Information Solubility in water: soluble. Other solubilities: miscible with alcohol, dimethylformamide,, chloroform, ether and most oils
Formula Weight 58.08
Percent Purity 99.8%
Grade HPLC
Chemical Name or Material Acetone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'