missing translation for 'onlineSavingsMsg'
Learn More

Acetone, for HPLC

Catalog Number p-7072774
30.94 EUR valid until 2024-12-31
Use promo code "21898" to get your promotional price.


30.94 EUR valid until 2024-12-31
Use promo code "21905" to get your promotional price.


30.94 EUR valid until 2024-03-30
Use promo code "21663" to get your promotional price.


35.65 EUR valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 L
2.5 L
67-64-1
67-64-1

C3H6O, CAS Number-67-64-1, 2.5L, 56 deg.C, Colorless, 58.08g/mol, -95 deg.C, 8765, Amber glass bottle, Liquid, 247mbar at 20 deg.C, 0.32 mPaS at 20 deg.C

TRUSTED_SUSTAINABILITY
pH 7
Melting Point -95°C
Color Colorless
Boiling Point 56°C
Quantity 1 L
Packaging Amber Glass Bottle
Physical Form Liquid
Molecular Formula C3H6O
MDL Number 8765
Vapor Pressure 247mbar at 20°C
Viscosity 0.32 mPaS at 20°C
Formula Weight 58.08g/mol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'