missing translation for 'onlineSavingsMsg'
Learn More

Acetone, Semiconductor Grade, 99.5%, Thermo Scientific Chemicals

Catalog Number p-3256840
Click to view available options
Quantity:
1 L
4 x 1 L
67-64-1
C3H6O
58.08
MFCD00008765
CSCPPACGZOOCGX-UHFFFAOYSA-N
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
180
CHEBI:15347
propan-2-one
CC(C)=O
67-64-1
C3H6O
58.08
MFCD00008765
CSCPPACGZOOCGX-UHFFFAOYSA-N
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
180
CHEBI:15347
propan-2-one
CC(C)=O

Maximum level of impurities: Color (APHA) 10, Evaporation residue 5ppm, Acidity 0.3μeq/g, Alkalinity 0.5μeq/g, H2O 0.5%, Solubility in H2O P.T., Cl 0.2ppm, PO4 0.1ppm

Acetone, semiconductor grade is suitable for use in electronics and circuit board manufacturing. Its luminescence intensity is dependent upon the solution components and the absorption of UV light induces photolysis and production of radials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point °C to 94°C
Color Colorless
Appearance Colorless liquid
Density 0.791
Boiling Point 56°C
Quantity 1 L
Flash Point -17°C (1°F)
Ignition Point 465°C (869°F)
Odor Characteristic
Physical Form Liquid
Refractive Index 1.359
Assay Percent Range 99.5%
Vapor Pressure 247 hPa (185mm Hg) at 20°C
Beilstein 63580
Solubility Information Miscible with water,benzene,alcohol,dimethylformamide,ether,chloroform and most oils.
Formula Weight 58.08
Grade Semiconductor
Chemical Name or Material Acetone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'