missing translation for 'onlineSavingsMsg'
Learn More

Acetone, Spectrophotometric Grade, 99.5+%, Thermo Scientific Chemicals

Catalog Number p-3256841
Click to view available options
Quantity:
1 L
4 x 1 L
This item is not returnable. View return policy
67-64-1
C3H6O
58.08
MFCD00008765
CSCPPACGZOOCGX-UHFFFAOYSA-N
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
180
CHEBI:15347
propan-2-one
CC(C)=O
This item is not returnable. View return policy
67-64-1
C3H6O
58.08
MFCD00008765
CSCPPACGZOOCGX-UHFFFAOYSA-N
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
180
CHEBI:15347
propan-2-one
CC(C)=O

Acetone is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point °C to 94°C
Color Colorless
Appearance Colorless liquid
Density 0.791 g/cm3 at 20°C
Boiling Point 56°C
Quantity 1 L
Flash Point -17°C (1°F)
Odor Characteristic
Physical Form Liquid
Refractive Index 1.359
Assay Percent Range >99.5%
Vapor Pressure 247 hPa (185mm Hg) at 20°C
Beilstein 635680
Solubility Information Miscible with water,benzene,alcohol,dimethylformamide,ether,chloroform and most oils.
Formula Weight 58.08
Grade Spectrophotometric
Chemical Name or Material Acetone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'