missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, 99.5%, for analysis, Thermo Scientific Chemicals

Catalog Number p-3779939
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal Drum
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
75-05-8
C2H3N
41.053
MFCD00001878
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
6342
CHEBI:38472
acetonitrile
CC#N
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
75-05-8
C2H3N
41.053
MFCD00001878
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
6342
CHEBI:38472
acetonitrile
CC#N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY
Melting Point -46.0°C
CAS Min % 99.4
CAS Max % 100.0
Density 0.7810g/mL
Boiling Point 81.0°C to 82.0°C
Quantity 1 L
Flash Point 2°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3430 to 1.3450
Assay Percent Range 99.5%
Residue after Evaporation 0.005% max.
Specific Gravity 0.781
Linear Formula CH3CN
Viscosity 0.36 mPa.s (20°C)
Beilstein 02, 183
Water 0.1% max. (Coulometric)
Fieser 02,13; 12,450; 13,156; 15,1
Merck Index 15, 69
Solubility Information Solubility in water: miscible. Other solubilities: soluble in alcohol, ethyl acetate, acetone, ether
Formula Weight 41.05
Percent Purity 99.5%
Grade Analytical
Chemical Name or Material Acetonitrile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'