missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, For pesticide residue analysis, Honeywell Riedel-de Haën™

Catalog Number p-7072098
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
This item is not returnable. View return policy
75-05-8
C2H3N
MFCD00001878
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
CHEBI:38472
acetonitrile
CC#N
This item is not returnable. View return policy
75-05-8
C2H3N
MFCD00001878
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
CHEBI:38472
acetonitrile
CC#N

for pesticide residue analysis

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.786g/mL at 25°C
Quantity 1 L
Explosion Limit 0.16
Flash Point 2°C (35.6°F)
Ignition Point 973°F
Linear Formula CH3CN
Vapor Pressure 72.8 mmHg (20°C)
UN Number UN1648
Beilstein 741857
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
Molecular Weight (g/mol) 41.053
Formula Weight 41.05g/mol
Percent Purity ≥99.9% (GC)
Grade CHROMASOLV™
Chemical Name or Material Acetonitrile
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'