missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile Anhydrous (DNA Synthesis), Fisher BioReagents

Catalog Number p-23824
Click to view available options
Quantity:
450 mL
4 L
Packaging:
Amber Glass
Glass Bottle
This item is not returnable. View return policy
75-05-8
C2H3N
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
6342
CHEBI:38472
acetonitrile
CC#N
This item is not returnable. View return policy
75-05-8
C2H3N
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
6342
CHEBI:38472
acetonitrile
CC#N
Acetonitrile Anhydrous is commonly used in DNA or peptide synthesis.
TRUSTED_SUSTAINABILITY
Melting Point -46°C
Absorbance 1 max. at 190nm, 0.15 max. at 195nm, 0.07 max. at 200nm, 0.05 max. at 205nm, 0.04 max. at 210nm, 0.02 max. at 220nm, 0.001 max. at 254nm
Color Colorless
Decomposition Information >500°C
Density 0.781g/cm³
Boiling Point 81°C
Quantity 450 mL
Packaging Amber Glass
Physical Form Liquid
Refractive Index 1.3405 to 1.3425 (at 25°C)
Assay Percent Range ≥99.9 %
Residue after Evaporation 1.0ppm max.
Titratable Acid 0.008mEq/g max.
Titratable Base 0.0006mEq/g max.
Vapor Pressure 97mbar at 20°C
UN Number 1648
Viscosity 0.36 cP at 20°C
Water 10ppm max.
Formula Weight 41.05
Percent Purity ≥99.9%
Grade DNA Synthesis
Chemical Name or Material Acetonitrile
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'