missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, HPLC for Gradient Analysis, meets analytical specification of Ph.Eur, Fisher Chemical™
137.17 EUR valid until 2024-12-31
Use promo code "21944" to get your promotional price.
139.00 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "21944" to get your promotional price.
139.00 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
Aluminum Can
Amber Glass Bottle
Plastic-coated Glass Bottle
Description
C2H3N, CAS Number-75-05-8, acetonitrile, cyanomethane, cyanure de methyl, ethanenitrile, ethyl nitrile, mecn, methanecarbonitrile, methane, cyano, methyl cyanide, methylkyanid, 2.5L, 81 deg.C, CHEBI:38472, Colorless, 41.05g/mol, WEVYAHXRMPXWCK-UHFFFAOYSA-N, acetonitrile, -46 deg.C, 1878, 41.053
Specifications
Melting Point | -46°C |
Color | Colorless |
Boiling Point | 81°C |
Quantity | 1 L |
Packaging | Amber Glass Bottle |
Vapor Pressure | 97mbar at 20°C |
Viscosity | 0.36cp at 20°C |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'