missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, Ultra Low Water, BakerDRY™, J.T. Baker™
Click to view available options
Quantity:
1 L
Packaging:
Narrow Mouth Amber Glass Bottle, w/ Septum
Specifications
Formula Weight | 41.05 |
Grade | BakerDRY™ |
Quantity | 1 L |
Flash Point | 2.00°C |
Packaging | Narrow Mouth Amber Glass Bottle, w/ Septum |
Specific Gravity | 1L = 0.79kg |
Chemical Name or Material | Acetonitrile, Ultra Low Water |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'