missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) AcroPak™ 20 Filters with Supor™ EKV Membrane
AcroPak 20 Filters are designed to add convenience to small- and medium-volume filtration (sterile and non-sterile).
88.60 € - 97.35 €
Specifications
Material (Membrane) | Hydrophilic PES |
---|---|
Pore Size | 0.2 μm |
Filtration Area | 20 cm2 |
Outlet Style | Stepped Hose Barb |
Quantity | 3/Pk. |
Product Code | Brand | Sterility | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterility | Price | Quantity & Availability | |||||
17164231
|
Cytiva (formerly Pall Lab)
12246 |
Non-sterile |
88.60 €
Pack of 3 |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17174231
|
Cytiva (formerly Pall Lab)
12247 |
Sterile |
97.35 €
Pack of 3 |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Supor membrane has high flow rates and throughputs, and is ideal for solutions where low protein binding is required. Not recommended for use with some ketones
- Efficiently processes up to 2 L of chemical and biological fluids
- Sterilization by gamma irradiation eliminates potential contamination by EtO residuals
- Upstream vent prevents vapor lock
- Built-in prefilter extends filter life when particulate-laden solutions such as serum-containing media are processed
Specifications
Hydrophilic PES | |
20 cm2 | |
3/Pk. |
0.2 μm | |
Stepped Hose Barb |