Learn More
Cytiva (formerly Pall Lab) AcroPrep™ 96 Filter Plates, 1 mL
Optimized for a variety of applications requiring larger per well volume processing.
Brand:  Cytiva (formerly Pall Lab) 5053
Additional Details : Weight : 0.65550kg
 View more versions of this product
Product Code. 17154461
  • 373.00 € / Pack of 5
Estimated Shipment: 23-08-2024
to see stock.
Add to basket
Description
Description
  • Plates are constructed of chemically resistant, biologically-inert polypropylene
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
Specifications
Glass Fiber (Borosilicate Glass Without Binder), Bio-Inert (Modified Low Protein-Binding Nylon)
Non-sterile
900 μL (Sample)
3.0/0.2 μm
5/Pk.