missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acrylamide, +99%, for biochemistry, electrophoresis grade

Catalog Number 10711441
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
This item is not returnable. View return policy
79-06-1
C3H5NO
71.08
MFCD00008032
HRPVXLWXLXDGHG-UHFFFAOYSA-N
Ethylenecarboxamide, 2-Propenamide
6579
CHEBI:28619
prop-2-enamide
NC(=O)C=C
This item is not returnable. View return policy
79-06-1
C3H5NO
71.08
MFCD00008032
HRPVXLWXLXDGHG-UHFFFAOYSA-N
Ethylenecarboxamide, 2-Propenamide
6579
CHEBI:28619
prop-2-enamide
NC(=O)C=C

Acrylamide, CAS # 79-06-1, is an organic amide compound used as a monomer in the synthesis of polyacrylamides.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Acrylamide is an organic amide compound mainly used to as a monomer in the synthesis of polyacrylamide
• Acrylamide is formed in foods containing carbohydrates in its composition, mainly when these foods are fried, grilled or baked

Applications

• In laboratory experiments acrylamide participates in chemical reactions as an intermediate in the preparation of N-methylol acrylamide and N-butoxyacrylamide compounds
• For molecular usage, this compound can be used as a carrier and is involved in the precipitation of small amounts of DNA
• In molecular experiments it is used in gel electrophoresis, including western blot, and SDS-PAGE, to separate proteins and macromolecules based on their size and charge

TRUSTED_SUSTAINABILITY
Physical Form Crystals or Crystalline Powder
Acrylic Acid 0.002% max.
Boiling Point 125.0°C (25.0 mmHg)
CAS Min % 99.0
CAS Max % 100.0
Melting Point 82.0°C to 86.0°C
Conductivity 5 umhos (40% max. aqueous solution)
Density 1.1220g/mL
Color White
pH 5.5 to 6.5 (10% soln., 0.1 M NaCl, 25°C)
Quantity 500 g
Assay Percent Range 99+%
Flash Point 138°C
Linear Formula H2C=CHCONH2
Beilstein 02, 400
Merck Index 15, 120
Infrared Spectrum Authentic
Solubility Information Solubility in water: 21.5g/L (30°C). Other solubilities: solubilities in g/100mL solvent (30°C):, methanol 155; ethanol 86.2; acetone 63.1;, ethyl acetate 12.6; chloroform 2.66;, benzene 0.346; heptane 0.0068, soluble in ether
Formula Weight 71.08
Percent Purity ≥99%
Grade Biochemical
Packaging Plastic Bottle
Chemical Name or Material Acrylamide, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'