Learn More
Eppendorf™ Adapter for S-4x1000 Rotor High-Capacity Bucket
Facilitates the use of specific sizes, shapes, and numbers of vessels in S-4x1000 rotor high-capacity buckets.
Brand: Eppendorf™ 5920724002
Additional Details : Weight : 0.75600kg
Description
- Suitable for S-4x1000 rotor high-capacity buckets
- Accommodates selected array of tubes or bottles
Specifications
Round Bottom (FACS) | |
12 mm | |
S-4X1000 rotor high-capacity buckets | |
52 |
52 x 5 mL Round Bottom Tubes (FACS) | |
106 mm | |
52 | |
2/Pk. |