missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Adapters For Fixed-Angle Rotor
Add flexibility to Eppendorf Centrifuges with these rotor adapters
Brand: Eppendorf™ 5804773004
Code : 99
Additional Details : Weight : 0.08000kg
Description
- Adapters
- For Eppendorf™ Fixed Angle Rotors
Specifications
Adapter | |
Fixed angle rotor | |
6 |
20 to 30 mL | |
26 mm | |
2 Pack |