missing translation for 'onlineSavingsMsg'
Learn More

Manutan Add-on bay for heavy duty steel shelving, 2000mm x 1250mm x 500mm

Brand:  Manutan 787M035

Code : Z3

Additional Details : Weight : 1.00000kg

Product Code. 12829088

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Create a run of shelving as long as you require. Heavy duty boltless shelving that stores up to 475kg per shelf. Versatile add-on shelving system with simple assembly - easy 'click-in' shelves. Choice of tubular or steel shelves with hard-wearing, powder-coated painted finish.
  • Five shelves
  • Extension bay with 5 steel shelves
  • Shelf capacity UDL: 460kg
  • Dimensions (H x W x D): 2000mm x 1260mm x 500mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers