Learn More
Epredia™ Adhesion Excell Slides
SuperFrost Excell slides are especially useful in applications involving high pH Heat Induced Epitope Retrieval (HIER), epon & methyl methacrylate sections, Laser Capture Microdissection (LCM) and cell culture.
Brand: Epredia™ J5800AMNZ
Code : A0
Additional Details : Weight : 0.33300kg
Description
- No time wasted on slide-coating proceduress
- No money wasted on expensive adhesives
- Reliable and uniform coating
- Minimal sample loss
- Reliable and safe adhesive action
Specifications
75 mm | |
25 mm | |
White |