missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ Adhesive Labels
Transparent, punched labels for use with wash bottles and plastic or glass reagent bottles
90.20 €
Specifications
Description | Transparent with colored text |
---|---|
Quantity | 50/Pk. |
Dimensions (LxW) | 130 x 35 mm |
Product Code | Brand | Color | Display Text | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Display Text | Price | Quantity & Availability | ||||
15895517
|
Kartell™
0501600 |
Black | Chloroform |
90.20 €
Pack of 50 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
15835517
|
Kartell™
0501000 |
Blue | Dist. Water |
90.20 €
Pack of 50 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
15885517
|
Kartell™
0501500 |
Green | Ethanol |
90.20 €
Pack of 50 |
In Stock
Log in to see stock. |
||||
15875517
|
Kartell™
0501400 |
Orange | Ethyl Acetate |
90.20 €
Pack of 50 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
15855517
|
Kartell™
0501200 |
Black | Methanol |
90.20 €
Pack of 50 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
15805527
|
Kartell™
0501800 |
Black | Methylene Cloride |
90.20 €
Pack of 50 |
Estimated Shipment: 28-06-2024
Log in to see stock. |
||||
Description
Specifications
Transparent with colored text | |
50/Pk. | |
130 x 35 mm |