missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Adjuvant Peptide

For cellular and molecular biology applications

Brand:  Alfa Aesar™ J62385.M¬

Code : NEW

Additional Details : CAS Number : 53678-77-6 Weight : 0.00001kg

 View more versions of this product

Product Code. 15414779

  • 27.00 € / 1mg


Estimated Shipment date
from Supplier 31-10-2018
Add to basket

Specifications

Specifications

492.48(anhy)
1mg
Powder
RUO
Muramyl dipeptide
HPLC
Used in immunization to replace more complex proteins. Inhibits the HIV replication in CD4+ H9 lymphocytes
N-Acetylmuramyl-L-alanyl-D-isoglutamine
2° to 8°C
Adjuvant Peptide
Videos
SDS

For Research Use Only