Learn More
Axygen™ Aerosol Filter Tips, Sterile
Prevent cross-contamination with robotic barrier tips. Axygen™ Aerosol Filter Tips, Sterile are e-beam irradiated to prevent product degradation and are free of leachable additives. Available in a wide variety of tip styles.
Brand:  Axygen™ TF-200-L-R-S
Code : 66
Additional Details : Weight : 8.20000kg
 View more versions of this product
Product Code. 12736739
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Technologically advanced filter barrier pipette tips
  • Ideal for DNA amplification and microbiological applications where trace contamination can invalidate results
  • Filter allows free passage of air while preventing cross-contamination by blocking the passage of aerosols or liquids
  • Nonpyrogenic, clear polyethylene filters are free from DNA inhibitors and leachable additives
  • Every filter is inserted into the tip robotically to ensure freedom from nuclease contamination
  • Packaged in sealed-bottom racks with lids to prevent contamination
  • Wide variety of tip styles available in a range of packaging quantities:
    • Maxymum Recovery™ technology provides excellent accuracy and prevents wastage of precious samples
    • Wide bore tips are ideal for pipetting viscous materials, genomic DNA or delicate cell cultures
Specifications
Specifications
1 to 200 μL
Sterile
Autoclavable
Rack
Gilson™ Pipetman™ P200
Filtered
Clear
DNase, RNase, and Pyrogen Free
Maxymum Recovery
Polypropylene