Learn More
Terumo™ Agani Hypodermic Needles
Used in conjunction with Terumo&trade Polypropylene Syringes. Terumo™ Agani Hypodermic Needles are triple-beveled and silicone lubricated for easy insertion.
5.30 €
Specifications
Material Stainless steel
Needle Point Style Triple-beveled, 11°
View More Specs
Products 7
Product Code Brand Needle Gauge Diameter (Metric) Outer Length (Metric) Needle Price Quantity & Availability  
Product Code Brand Needle Gauge Diameter (Metric) Outer Length (Metric) Needle Price Quantity & Availability  
15301557 Terumo™
8AN2613R1
26 ga. 0.45mm 13mm 5.30 €
Pack of 100
Estimated Shipment
06-02-2019
Add to basket
15321547 Terumo™
8AN2238R1
22 ga. 0.7mm 38mm 5.30 €
Pack of 100
Estimated Shipment
06-02-2019
Add to basket
15341547 Terumo™
8AN2325R1
23 ga. 0.6mm 25mm 5.30 €
Pack of 100
Estimated Shipment
06-02-2019
Add to basket
15341557 Terumo™
8AN2719R1
27 ga. 0.4mm 19mm 5.30 €
Pack of 100
UK Stock 10
Add to basket
15361537 Terumo™
8AN2038R1
20 ga. 0.9mm 38mm 5.30 €
Pack of 100
UK Stock 6
Add to basket
15371547 Terumo™
8AN2516R1
25 ga. 0.5mm 16mm 5.30 €
Pack of 100
UK Stock 8
Add to basket
15391537 Terumo™
8AN2138R1
21 ga. 0.8mm 5.30 €
Pack of 100
UK Stock 7
Add to basket
Description
Description
  • Stainless steel
  • Triple-beveled points
  • Silicone lubrication
Specifications
Specifications
Stainless steel
Triple-beveled, 11°