missing translation for 'onlineSavingsMsg'
Learn More

Terumo™ Agani Hypodermic Needles

Used in conjunction with Terumo™ Polypropylene Syringes. Terumo™ Agani Hypodermic Needles are triple-beveled and silicone lubricated for easy insertion.

Brand:  Terumo™ 8AN2238R1

Additional Details : Weight : 0.14000kg

 View more versions of this product

Product Code. 15321547

  • 5.35 € / Pack of 100

Estimated Shipment
22-06-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Stainless steel
  • Triple-beveled points
  • Silicone lubrication
Specifications

Specifications

Black
0.7 mm
Stainless Steel
22 G
38 mm
11° Triple-beveled
Videos
Documents