Learn More
Terumo™ Agani™ Single-use Sterile Hypodermic Needles
Make procedures as painless as possible with extremely sharp and reliable stainless steel needles. Terumo™ Agani™ Single-use Sterile Hypodermic Needles feature a unique, latex-free double bevel design for maximum comfort.
Brand:  Terumo™ 8AN2138R1
Additional Details : Weight : 0.12600kg
 View more versions of this product
Product Code. 15391537
  • 7.84 € / Pack of 100
In Stock
to see stock.
Description
Description
  • Constructed from polypropylene, stainless steel, epoxy resin and dimethyl silicone oil
  • All needles are supplied sterile wrapped in individual, tamper proof peel pouches
  • Simple color coded system allows easy identification of required sizes
  • Use with a wide range of Sterile Syringes
  • Needles are sterilized with ethylene oxide to a Sterility Assurance Level of at least 10-6 Class IIa, sterile product following MDD 93/42/EEC
  • CE marked and latex free
  • Supplied in boxes of 100 needles
Specifications
Specifications
Stainless Steel
21 G
38 mm
11° Triple Bevel
Green
0.8 mm
Sterile Hypodermic Needle for Single-Use
Sterile