Learn More
Molecular Probes™ Alexa Fluor™ 488 Signal-Amplification Kit for Mouse Antibodies
Alexa Fluor™ 488 Signal-Amplification Kit for Mouse Antibodies
Brand:  Molecular Probes™ A11054
Additional Details : Weight : 0.00450kg
Product Code. 10636543
  • 266.00 € / Each
Estimated Shipment: 10-05-2024
to see stock.
Description
Description
For sensitive detection of mouse primary antibodies, try our Alexa Fluor™ 488 Signal-Amplification Kit, which contains Alexa Fluor 488 rabbit anti—mouse IgG and Alexa Fluor 488 goat anti—rabbit IgG conjugates. This kit takes advantage of the intense, photostable fluorescence of Alexa Fluor 488 antibody conjugates and can be used to amplify the signals derived from mouse-derived antibodies. The kit contains a detailed protocol and sufficient reagents for ∼60 assays by flow cytometry or imaging. For added convenience, the kit is designed for use with filters and settings appropriate for fluorescein.
Specifications
Specifications
Store in refrigerator (2–8°C) and protect from light.
Room Temperature
Alexa Fluor™ 488
Alexa Fluor™