Learn More
Thermo Scientific Chemicals Ammonium, Quant|r Test Strips
Brand:  Thermo Scientific Chemicals 023267.QA
Code : NEW
Additional Details : Weight : 0.01000kg Transport : UN number : 1824 Chem class : 8 Pack group : III
Product Code. 15481117
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alert:
H314; Causes severe skin burns and eye damage. P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a; Do not breathe dust/fume/gas/mist/vapours/spray. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations.
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Fully miscible
  • Liquid form
Specifications
Specifications
Liquid
Reagents and strips to measure 10-400ppm NH4+1 in aqueous solution
Ambient
Fully Miscible
Product Suggestions
Product Suggestions
Videos
SDS Documents