missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Disposable IR cards, 5x10cm, KBr window, 15mm aperture
Cardboard card that fits into the holder of a spectrophotometer
132.00 € - 334.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15446575
|
Thermo Scientific Chemicals
043936.KJ |
5 Pk. |
132.00 €
Pack of 5 |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15456575
|
Thermo Scientific Chemicals
043936.KM |
25 Pk. |
334.00 €
Pack of 25 |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Dimensiosn (L × H): 50 × 100mm (1.97 × 3.94 in.)
- Disposable cardboard cards
- Aperture: 15mm (0.591 in.)
- KBr window
RUO – Research Use Only