Learn More
Formaldehyde, Quant™ Test Strips, Thermo Scientific Chemicals
Brand:  Thermo Scientific Alfa Aesar 023277.QA
Code : NEW
Additional Details : Weight : 0.01000kg Transport : UN number : 1824 Chem class : 8 Pack group : III
Product Code. 15461127
  • 263.00 € / Each
Estimated Shipment: 06-06-2024
to see stock.
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
1 Kit
Measures 0 to 100ppm HCHO in aqueous solution
Formaldehyde test strip
Product Suggestions
Product Suggestions
Videos
SDS Documents