Learn More
Thermo Scientific Chemicals Sapphire substrate, 50.8mm dia x 0.332mm thick, EPI polished two sides, C-axis, HEMCOR single crystal
263.00 € - 1141.00 €
Product Code | Brand | Quantity | Specimen Size | Thickness | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Specimen Size | Thickness | Price | Quantity & Availability | |||
15463925
|
Thermo Scientific Chemicals
045020.KT |
1 Ea. | 50.8mm dia. x 0.332 mm thick | 0.332 mm |
263.00 €
Each |
In Stock
Log in to see stock. |
|||
15473925
|
Thermo Scientific Chemicals
045020.KU |
5 Ea. | 50.8mm dia. x 0.332 mm thick | 0.332 mm |
1141.00 €
Pack of 5 |
Estimated Shipment: 20-05-2024
Log in to see stock. |
|||