missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Sapphire Substrate, EPI Polished Two Sides, C-axis, HEMCOR Single Crystal

Brand:  Alfa Aesar™ 045020.KU

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15473925

  • 1013.00 € / Pack of 5

Estimated Shipment date
from Supplier 17-09-2019

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

Sapphire substrate, EPI polished two sides
50.8mm dia. x 0.332mm thick
EPI polished two sides
5 Each
50.8mm
0.332mm
Videos
SDS Documents