missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan All fixed mesh sided metal pallet, 1220mm x 915mm, 500kg capacity
Brand: Manutan A337954
Code : Z3
Additional Details : Weight : 1.00000kg
Description
A range of welded steel pallets that are ideal for bulk storage, saving valuable floor space. Mesh steel sides and sheet steel decks, all with 115mm ground clearance. Stackable up to 5 high.- Load height: 760mm
- Dimensions (W x D): 1220mm x 915mm