Learn More
Allihn Condenser with Ground Cone and Socket
Ideal for reflux applications.
Brand: Quickfit CX7/44/SC
Code : NEW
Additional Details : Weight : 0.43000kg
Description
- Inner bulb sections provide increased cooling surface
- With detachable plastic screwthread connectors for easy and safe attachment of flexible tubing
Specifications
29/32 | |
550 mm | |
Borosilicate Glass |
Allihn Condenser | |
1 Each |