missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha-1-Antitrypsin Mouse, Clone: AAT/1378, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 485.00 €
Specifications
Antigen | Alpha-1-Antitrypsin |
---|---|
Clone | AAT/1378 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15960638
|
EnquireBio
5265-MSM1-P1 |
100 μg |
485.00 €
100µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950638
|
EnquireBio
5265-MSM1-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Alpha-1-Antitrypsin | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
5265 | |
Recombinant human Alpha-1-Antitrypsin (AAT) protein fragment | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
54kDa |
AAT/1378 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_001121173 | |
SERPINA1 | |
IgG1 | |
Protein A/G Purified | |
Cytoplasmic | |
HepG2 Cells. Tonsil, Hepatocellular Carcinoma or Histiocytoma. |