missing translation for 'onlineSavingsMsg'
Learn More

alpha-Benzoin oxime, 98%, ACROS Organics™

Brand:  Acros Organics 105535000

Code : 29

Additional Details : CAS Number : 441-38-3 Weight : 0.50000kg

Packaging Glass bottle
Quantity 500g

 View more versions of this product

Product Code. 11955651

  • 281.00 € / 500g

In stock, ready to ship: 2

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

441-38-3
227.26
2-hydroxy-1,2-diphenylethanone oxime, e,alphar-benzoinoxime, 2r-2-hydroxy-1,2-diphenylethanone oxime, 1r,2z-2-hydroxyimino-1,2-diphenylethanol, 1r,2z-2-hydroxyimino-1,2-diphenyl-ethanol, unii-ptc1433kj8 component
(1R,2Z)-2-hydroxyimino-1,2-diphenylethanol
C14H13NO2
WAKHLWOJMHVUJC-JNINTMOKSA-N
7057888
C1=CC=C(C=C1)C(C(=NO)C2=CC=CC=C2)O

Specifications

α-Benzoin oxime
Glass bottle
150°C to 155°C
441-38-3
100.0
C14H13NO2
08,IV,1282
2-hydroxy-1,2-diphenylethanone oxime, e,alphar-benzoinoxime, 2r-2-hydroxy-1,2-diphenylethanone oxime, 1r,2z-2-hydroxyimino-1,2-diphenylethanol, 1r,2z-2-hydroxyimino-1,2-diphenyl-ethanol, unii-ptc1433kj8 component
WAKHLWOJMHVUJC-JNINTMOKSA-N
(1R,2Z)-2-hydroxyimino-1,2-diphenylethanol
7057888
Crystalline Powder
Authentic
White
500g
97.5
97.5% min. (HPLC)
C6H5CH(OH)C(=NOH)C6H5
15,1096
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,aq. ammonium hydroxide
C1=CC=C(C=C1)C(C(=NO)C2=CC=CC=C2)O
227.26
227.26
98%
Videos
SDS Documents
Certificates