missing translation for 'onlineSavingsMsg'
Learn More

alpha-Benzoin oxime, 98%, Thermo Scientific Chemicals

Catalog Number p-4229078
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
441-38-3
C14H13NO2
227.26
MFCD00004501
WAKHLWOJMHVUJC-FYWRMAATNA-N
2-hydroxy-1,2-diphenylethanone oxime, e,alphar-benzoinoxime, 2r-2-hydroxy-1,2-diphenylethanone oxime, 1r,2z-2-hydroxyimino-1,2-diphenylethanol, 1r,2z-2-hydroxyimino-1,2-diphenyl-ethanol, unii-ptc1433kj8 component
7057888
(1R,2Z)-2-hydroxyimino-1,2-diphenylethanol
O\N=C(\C(O)C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
441-38-3
C14H13NO2
227.26
MFCD00004501
WAKHLWOJMHVUJC-FYWRMAATNA-N
2-hydroxy-1,2-diphenylethanone oxime, e,alphar-benzoinoxime, 2r-2-hydroxy-1,2-diphenylethanone oxime, 1r,2z-2-hydroxyimino-1,2-diphenylethanol, 1r,2z-2-hydroxyimino-1,2-diphenyl-ethanol, unii-ptc1433kj8 component
7057888
(1R,2Z)-2-hydroxyimino-1,2-diphenylethanol
O\N=C(\C(O)C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 150°C to 155°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH(OH)C(=NOH)C6H5
Quantity 25 g
Beilstein 08,IV,1282
Merck Index 15,1096
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol,aq. ammonium hydroxide
Formula Weight 227.26
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material α-Benzoin oxime
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'