missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ ALUGRAM™ Xtra SIL G Aluminum Sheets

Stable in almost all organic solvents and in purely aqueous eluents. Macherey-Nagel™ ALUGRAM™ Xtra SIL G Aluminum Sheets are unmodified standard silica layers on aluminum for use in TLC. Feature outstanding wettability and improved cutting properties.

Brand:  Macherey-Nagel™ 818261

Code : 50

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12750763

  • 59.00 € / Pack of 50
Estimated Shipment: 24-05-2024
to see stock.
Add to basket

Description

Description

ALUGRAM™ Xtra SIL G Aluminum Sheets feature unmodified, standard-grade silica 60 layers without a fluorescent indicator.

  • Outstanding wettability for precise colorization results, even with 100% aqueous detection reagents
  • Excellent separation efficiency and reproducibility from lot to lot
  • Easy and reliable cutting due to an optimized binder system, no flaking of silica
  • Stable in almost all organic solvents and resistant towards aggressive visualization reagents
  • Highly polymeric product is also completely stable in purely aqueous eluents
Specifications

Specifications

Alugram Aluminum Sheets with IUInmodified Silica 60 Layers
60 Å
Aluminum Sheet
∼500 m2/g
5 x 10 cm
5 to 17 μm
50 Pk.
0.75 mL/g
0.20 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers