missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Aluminum Alloy Rods
Approximately 13mm in diameter.
3.50 € - 7.15 €
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10265394
|
Fisherbrand™
|
100 mm |
3.50 €
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
10133784
|
Fisherbrand™
|
200 mm |
4.40 €
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
10624055
|
Fisherbrand™
|
400 mm |
7.15 €
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
Description
- Available in a range of lengths
- Excellent for building scaffolding in the laboratory
Specifications
Aluminum Alloy | |
Rod |