missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum, technical, foil, ≈0.1mm thick, SLR, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
Specifications
Color | Gray-Silver |
Melting Point | 660°C |
Boiling Point | 2327°C |
Quantity | 250 g |
Formula Weight | 26.97g/mol |
Packaging | Plastic container |
Form | Solid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'