missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Copper spheres, alloy 2017, 19.0mm (0.75 in.) dia., Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar 042036.KG

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15406425

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Aluminum Copper spheres, Alloy 2017
Al:Cu:Mn:Mg; 94.8:4:0.7:0.5 wt%
1 Pc.
19.0mm (0.75 in.) dia.
MFCD00213972
Odorless
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Special Offers

Special Offers