missing translation for 'onlineSavingsMsg'
Learn More

Statebourne Cryogenics™ BioTrek™ Series Cryogenic Dry Shippers

Designed to transport large or small biological specimens. Statebourne Cryogenics™ BioTrek™ Series Cryogenic Dry Shippers are supplied with neck plugs.

POA

Specifications

For Use With (Application) Storage of liquid nitrogen
Material Aluminum
View More Specs

Description

Description

  • Offers storage times of up to 18 days, depending on model
  • 3L model includes single canister, 10L model does not include canister system
  • Special internal absorbent material can be charged with liquid nitrogen in minutes to retain cryogenic temperatures for the whole holding time
Specifications

Specifications

Storage of liquid nitrogen
Aluminum