Learn More
Aluminium hydroxide, extra pure, powder, ACROS Organics™
Brand: Acros Organics 219130100
Code : 29
Additional Details : CAS Number : 21645-51-2 Weight : 10.05000kg
Packaging | Plastic drum |
---|---|
Quantity | 10kg |
Chemical Identifiers
21645-51-2 | |
AlH3O3 | |
78 |
Specifications
Aluminium hydroxide | |
AlH3O3 | |
15,338 | |
78 | |
Powder | |
Extra Pure | |
Plastic drum | |
White | |
10kg |
21645-51-2 | |
Al(OH)3 | |
Solubility in water: insoluble | |
78 | |
≥63.5% (Al2O3) | |
36.5% max. | |
0.2% max. | |
8.5 to 10 (5% aq. suspension) |
Safety and Handling
- Aluminium hydroxide
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.